Alfa Aesar™ Cortistatin-17, Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66418EXD
 View more versions of this product
Catalog No. AAJ66418EXD
Specifications
Specifications
2151.51
Human
Solid
RUO
Cortistatin-17, human
HPLC
0.5mg
Asp-Arg-Met-Pro-Cys-Arg-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys (Disulfide bridge: Cys5-Cys16)
-30° to -10°C