Please login to your online account to display your discounted pricing

Alfa Aesar™ Cortistatin-17, Human

For cellular and molecular biology applications

Specifications

Product Type Cortistatin-17, human
Cross Reactivity Human
Molecular Weight 2151.51
Storage Requirements -30° to -10°C
Sequence Asp-Arg-Met-Pro-Cys-Arg-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys (Disulfide bridge: Cys5-Cys16)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66418EXD

 
alfa aesar™
J66418EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66418MCR

 
alfa aesar™
J66418MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Cortistatin-17, human
Cross Reactivity Human
Molecular Weight 2151.51
Storage Requirements -30° to -10°C
Sequence Asp-Arg-Met-Pro-Cys-Arg-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys (Disulfide bridge: Cys5-Cys16)
Format Solid
Regulatory Status RUO
Purification Method HPLC