Please login to your online account to display your discounted pricing

Alfa Aesar™ Cortistatin 14

For cellular and molecular biology applications

Specifications

Product Type Cortistatin 14
Molecular Weight 1721.10
Storage Requirements -30° to -10°C
Sequence Pro-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys (Disulfide bridge: Cys2-Cys13)
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66255EXD

 
alfa aesar™
J66255EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66255MCR

 
alfa aesar™
J66255MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Cortistatin 14
Molecular Weight 1721.10
Storage Requirements -30° to -10°C
Sequence Pro-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys (Disulfide bridge: Cys2-Cys13)
Format Solid
Regulatory Status RUO
Purification Method HPLC