Please login to your online account to display your discounted pricing

Cortisol Parameter Assay Kit, R&D Systems

$376.73 - $1,808.04

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
For Use With (Application) For the quantitative determination of cortisol concentrations in cell culture supernates, serum, saliva and urine
Sensitivity 0.111ng/mL
Species Multi-species
View More Specs
Products
Catalog Number Mfr. No. Sufficient For Product Type Quantity Price Quantity    

KGE008

 
r & d systems
RDSKGE008
1 x 96-well plate ELISA kit 1 kit Each for $376.73

SKGE008

 
r & d systems
RDSSKGE008
6 x 96-well plates ELISA kit 1 Pk. Each for $1,808.04
Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
For Use With (Application) For the quantitative determination of cortisol concentrations in cell culture supernates, serum, saliva and urine
Sensitivity 0.111ng/mL
Species Multi-species

Assay Type: Solid Phase Competitive ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (100μL), Saliva (100μL), Urine (100μL)
Sensitivity: 0.111ng/mL
Specificity: Measures Cortisol levels in various samples
Cross Reactivity: Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.