Please login to your online account to display your discounted pricing

Cortisol Parameter Assay Kit, R&D Systems


Manufacturer: r & d systems  RDSSKGE008

 View more versions of this product

Catalog No. SKGE008

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For the quantitative determination of cortisol concentrations in cell culture supernates, serum, saliva and urine
Product Type ELISA kit
Quantity 1 Pk.
Sensitivity 0.111ng/mL
Species Multi-species
Sufficient For 6 x 96-well plates
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.