missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coronene 95%, ACROS Organics™

Manufacturer:  Acros Organics 110515000

 View more versions of this product

Catalog No. AC110515000



Specifications

Specifications

Coronene
Glass bottle
500mg
191-07-1
C24H12
15, 2523
VPUGDVKSAQVFFS-UHFFFAOYSA-N
coronene
9115
300.35
≥94%
Authentic
Green to Yellow
95%
95%
MFCD00004134
hexabenzobenzene, dibenzo ghi,pqr perylene, superbenzene, unii-7yy0x5xt1w, ccris 908, 7yy0x5xt1w, dibenzo ghi,pqr perylene, 6 circulene, coronene, acmc-209eul
C1=CC2=C3C4=C1C=CC5=C4C6=C(C=C5)C=CC7=C6C3=C(C=C2)C=C7
300.36
CHEBI:29863
Crystalline Powder

Chemical Identifiers

191-07-1
300.36
VPUGDVKSAQVFFS-UHFFFAOYSA-N
9115
coronene
C24H12
MFCD00004134
hexabenzobenzene, dibenzo ghi,pqr perylene, superbenzene, unii-7yy0x5xt1w, ccris 908, 7yy0x5xt1w, dibenzo ghi,pqr perylene, 6 circulene, coronene, acmc-209eul
CHEBI:29863
C1=CC2=C3C4=C1C=CC5=C4C6=C(C=C5)C=CC7=C6C3=C(C=C2)C=C7
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-881-7

RTECSNumber : GM5400000

SDS Documents