Corning™ VISTA™ Petri Culture Dishes
Flat, clear dishes feature beaded edges for greater mechanical strength.
Manufacturer:  Corning™ 70165100EMD
 View more versions of this product
Catalog No. S95184A
Description
Description
Bead also provides a means to equally space the side walls of the bottom and cover, reducing the capillary action of the condensed moisture on the sides.
  • Manufactured from soda lime silica glass
  • Unaffected chemically or thermally in routine sterilizations
  • Withstand repeated sterilization — wet or dry
  • Bottoms have a triangular enamel reference point for serial dilutions
  • Covered dish is not airtight
Specifications
Specifications
Clear
95 mm
Beaded
Round
Education
Soda Lime Glass
127 mm
Documents