missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ VISTA™ Petri Culture Dishes
Flat, clear dishes feature beaded edges for greater mechanical strength.
Manufacturer: Corning™ 70165102EMD
Description
Bead also provides a means to equally space the side walls of the bottom and cover, reducing the capillary action of the condensed moisture on the sides.
- Manufactured from soda lime silica glass
- Unaffected chemically or thermally in routine sterilizations
- Withstand repeated sterilization — wet or dry
- Bottoms have a triangular enamel reference point for serial dilutions
- Covered dish is not airtight
Specifications
Clear | |
Round | |
22 mm |
Soda Lime Glass | |
Beaded | |
95 mm |