Please login to your online account to display your discounted pricing
Corning™ Vented Screw Caps for Falcon™ Flasks
Designed for use with Falcon 25-, 75-, and 175cm2 flasks
Manufacturer: corning™  C354639
 View more versions of this product
Catalog No. 08-774-205
  • / Case of 50 Each
Description & Specifications
Specifications
Product Type Cap, Vented
For Use With (Equipment) Falcon™ 175cm2 flasks
Includes 0.2μm Membrane vent
Sterile Yes
Closure Material Polyethylene
Closure Type Screw