Please login to your online account to display your discounted pricing
Corning™ Vented Screw Caps for Falcon™ Flasks
Designed for use with Falcon 25-, 75-, and 175cm2 flasks
Manufacturer: corning™  C354638
 View more versions of this product
Catalog No. 08-774-204
  • / Case of 100 Each
Description & Specifications
Specifications
Product Type Cap, Vented
Includes 0.2μm Membrane vent
Closure Type Screw
For Use With (Equipment) Falcon™ 75cm2 flasks
Closure Material Polyethylene
Sterile Yes