Corning™ Universal Fit Pipet Tips: Racked, Nonsterile
Designed to assure a reliable fit with major brand pipetters
Manufacturer: Corning™ C4863
Description
Designed to assure a reliable fit with major brand pipetters
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 µL universal fit tips are graduated at the 250 µL, 500 µL, and 1,000 µL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
1 to 200 μL | |
Natural | |
Round | |
Rack | |
Rack |
Non-sterile | |
Yes | |
PP (Polypropylene) | |
Universal |