Corning™ Universal Fit Hinged Rack Pipet Tips, Nonsterile
Hinged closure provides easy stacking and steam penetration during autoclaving
Manufacturer: Corning™ C4710
Description
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 µL universal fit tips are graduated at the 250 µL, 500 µL, and 1,000 µL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Order Info
Available sterile and nonsterile
Specifications
1 to 200 μL | |
Yellow | |
Round | |
Rack Insert | |
Hinged rack |
Non-sterile | |
Yes | |
Hinged closure provides easy stacking and steam penetration during autoclaving | |
Universal |