Corning™ Universal Fit Hinged Rack Pipet Tips, Nonsterile
Hinged closure provides easy stacking and steam penetration during autoclaving
Manufacturer:  Corning™ C4715
 View more versions of this product
Catalog No. 07-201-765
Description
Description
  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 µL universal fit tips are graduated at the 250 µL, 500 µL, and 1,000 µL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Order Info
Available sterile and nonsterile
Specifications
Specifications
100 to 1000 μL
Blue
Round
Rack Insert
Hinged rack
Non-sterile
Yes
Hinged closure provides easy stacking and steam penetration during autoclaving
Universal