missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Universal CellSTACK™ Caps

For filling, venting, and manipulation

Manufacturer:  Corning™ C3329

 View more versions of this product

Catalog No. 07-202-520



Description

Description

  • Choice of 33mm polyethylene, solid or vented
  • Sterilized by gamma irradiation

Specifications

Specifications

Orange
3.3 cm
Female MPC Polycarbonate with a 0.95cm I.D. coupling and a male MPC Polycarbonate end cap
Yes
Screw, Vented
HDPE
For filling, venting, and manipulation
Cap, Filling
Sterile
SDS
Documents