Corning™ Costar™ Ultra-Low Attachment Microplates
Featuring a covalently bound hydrogel layer that effectively inhibits cellular attachmment. Corning™ Costar™ Ultra-Low Attachment Microplates greatly reduce binding of attachment and serum proteins, minimizing cell attachment and spreading.
$1,051.21 - $1,233.72
Specifications
Culture Area 0.32cm2
Color Clear
Sterile Yes
Lid Yes
Material Polystyrene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Culture Area Diameter (Metric) Well No. of Wells Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Culture Area Diameter (Metric) Well No. of Wells Volume (Metric) Price Quantity  
07-200-602 Corning™
C3473
1.9cm2 1.56cm 24 3.4mL Case of 24 Each for $1,110.13
07-200-603 Corning™
C3474
0.32cm2 0.64cm 96 360μL Case of 24 Each for $1,233.72
07-200-601 Corning™
C3471
9.5cm2 3.48cm 6 16.8mL Case of 24 Each for $1,051.21
Specifications
Description
Biologically Inert Surface
  • Prevents stem cells from attachment-mediated differentiation
  • Prevents anchorage-dependent cells from dividing
  • Minimizes protein absorption, enzyme activation, and cellular activation
  • Noncytotoxic and nondegradable
Convenient Design
  • Made from optically clear polystyrene
  • Individual alphanumeric codes provide well identification
  • Uniform footprint aids stacking
  • Flat well shape
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
0.32cm2
Yes
Polystyrene
Clear
Yes