missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Two- and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks

Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Two-and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.

Manufacturer:  Corning™ C431255

 View more versions of this product

Catalog No. 10-126-42


 

Specifications

Description

Flask, Erlenmeyer; Corning; Polycarbonate; 2000mL; Nonbaffled; Flat and Vented cap; Disposable

 

  • Fernbach design increases surface area for gas exchange
  • Can be used for cultures or storing media
  • Vent caps supplied in every case of product for applications requiring sterile gas exchange
  • Available in baffled and nonbaffled bottom shape
  • Individually wrapped
  • Sterilized by gamma radiation and certified nonpyrogenic
  • RNase-/DNase-free
  • Autoclavable according to furnished instructions

 

Description & Specifications

Specifications

Non-baffled
Orange
Top style: Threaded
For shaker culture applications
Wide
2000mL
67.6 oz.
Polypropylene
Yes
100mL
Sterile
Polycarbonate