Please login to your online account to display your discounted pricing

Corning™ Two- and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks


Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Two-and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.

Manufacturer: corning™  C431252

 View more versions of this product

Catalog No. 10-126-38

  • / Case of 4 Each
 

Description & Specifications

Specifications

Closure Color Orange
For Use With (Application) For shaker culture applications
Closure Material Polypropylene
Sterile Yes
Capacity (English) 101.44 oz.
Capacity (Metric) 3000mL
Capacity 101.44 oz., 3000mL
Disposable Yes
Mouth Style Wide
Bottom Shape Non-baffled
Top Style Threaded
Material Polycarbonate

 

  • Fernbach design increases surface area for gas exchange
  • Can be used for cultures or storing media
  • Vent caps supplied in every case of product for applications requiring sterile gas exchange
  • Available in baffled and nonbaffled bottom shape
  • Individually wrapped
  • Sterilized by gamma radiation and certified nonpyrogenic
  • RNase-/DNase-free
  • Autoclavable according to furnished instructions