Please login to your online account to display your discounted pricing

Corning™ Transferrin, Human (Holo-form) Media Additive


Promotes the proliferation of MDCK cells in serum-free medium

Manufacturer: corning™  C354304

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. CB40304

  • / Each
 

Description & Specifications

Specifications

Quantity 1g
Storage Requirements 2° to 8°C
Solution Type Human Transferrin
Quantity 1g