missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates

Manufactured from ultrathin polycarbonate for fast, consistent heat transfer. Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.

Manufacturer:  Corning™ C6509

 View more versions of this product

Catalog No. 07-200-541



Description

Description

  • Low protein-binding surface prevents loss of sample
  • For oil-free operation, use polyethylene or aluminum sealing tape in conjunction with heated-lid thermal cyclers; also accommodates silicone reusable sealing mat
  • Withstands temperatures to 135°C without warping
  • Various well shapes fit popular thermal cyclers
  • Surface of DNA-BIND™ plate allows covalent attachment of ssDNA at 5« and 3« end
  • Nonsterile
Specifications

Specifications

Standard
Round
PCR
Polycarbonate
25/Cs.
Non-sterile
96
Without Lid
Clear
Applied Biosystems GeneAmp™ PCR System 9600b, Barnstead Thermolyne Amplitron II™, Techne™ Cyclogene, and Gene E with 96 x 0.2 mL block
25/Cs.
1/Pk.
Clear
8 x 12
SDS
Documents
Accessories