Please login to your online account to display your discounted pricing

Corning™ TC-Treated Culture Dishes


Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro.

Manufacturer: corning™  C430293

 View more versions of this product

Catalog No. 08-772-23

  • / Case of 480 Each
 

Description & Specifications

Specifications

Surface Area (Metric) 55cm2
Dimensions (L x W) 100 x 20mm
Sterile Yes
Diameter (Metric) 100mm
Surface Coating TC
Bottom Shape Round (U)
Material Polystyrene
Height (Metric) Exterior 20mm
Color Clear

  • Manufactured from optically-clear virgin polystyrene
  • Supplied with vents to provide consistent gas exchange
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling