Please login to your online account to display your discounted pricing

Corning™ T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Rat


Promotes polyclonal activation and IL-2 proliferation of most antigenically sensitized rat or mouse lymphoid T-cells

Manufacturer: corning™  C354116

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. CB-40116

  • / Each
 

Description & Specifications

Specifications

Product Type T-cell culture Supplement, Without ConA
Source Rat
Without Additives ConA
Storage Requirements 2° to 8°C
Concentration ≤0.5μg/mL
Format Max ConA Content: ≤ 0.5μg/mL
Quantity 100mL