Corning™ T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Rat
Promotes polyclonal activation and IL-2 proliferation of most antigenically sensitized rat or mouse lymphoid T-cells
$1,194.53 - $1,912.57
Specifications
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Products 2
Catalog Number Mfr. No. Product Type Format Price Quantity  
Catalog Number Mfr. No. Product Type Format Price Quantity  
CB-40116 View Documents Corning™
C354116
T-cell culture Supplement, Without ConA Max ConA Content: ≤ 0.5μg/mL Each for $1,912.57
CB-40115 View Documents Corning™
C354115
T-cell culture Supplement, With ConA Max ConA Content: 10μg/mL Each for $1,194.53
Description
Description
  • Conditioned medium (RPMI 1640) from ConA-stimulated rat splenocyte cultures containing 10% fetal bovine serum and 10mM HEPES
  • Medium with ConA supports high-titer HIV production by leukocytes
  • Medium without ConA (from washed rat splenocyte cultures) designed for applications that support proliferation of lectin-activated T-cells and single-cell cloning of antigen-specific T-cells from polyclonally activated primary cultures
  • Both undergo an appropriate bioassay, are filtered through a 0.2μm membrane and are tested for the absence of bacteria, fungi and Mycoplasma—guaranteeing a high-quality product
Order Info
Supplied in 100mL bottles; refrigerate as detailed on label
Specifications
Specifications
0.0
icon-pdf.png