Corning™ Surface Modified Stripwell™ Microplates
Four surface chemistries suit every type of assay
Manufacturer:  Corning™ C2506
 View more versions of this product
Catalog No. 07-200-585
Description
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Specifications
Specifications
Clear
Polystyrene
50/Cs.
1/Pk.
Non-sterile
96
Without Lid
Strips
In-vitro diagnostic assays
50/Cs.
DNA-Bind
360 μL
DNA-Bind
8 x 12
75 to 200 μL
Round