Corning™ Stripwell™ Microplates
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Manufacturer:  Corning™ C3913
 View more versions of this product
Catalog No. 07-200-593
Description
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Specifications
Specifications
Strips
75 to 200 μL
In-vitro diagnostic assays
100/Cs.
Medium Binding
360 μL
Round
96
Without Lid
White
Polystyrene
100/Cs.
25/Pk.
Non-sterile
Medium Binding
8 x 12