Corning™ Sterile Spatulas
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
$595.41 - $595.41
Specifications
Certifications/Compliance USP Class VI, non-pyrogenic, RNase and DNase-free Certified
Product Type Spatula
Disposable Yes
For Use With (Application) Molecular biology applications, lyophilized samples and contamination-free sampling
Material Polystyrene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Length (Metric) Price Quantity  
Catalog Number Mfr. No. Includes Length (Metric) Price Quantity  
14-245-95 Corning™
C3003
Tapered flat blade at one end and a spoon at the other end 24.9cm Case of 100 Each for $595.41
14-245-96 Corning™
C3004
Narrow small spoon at one end and a larger round spoon at the other end 23.4cm Case of 100 Each for $595.41
14-245-97 Corning™
C3005
Round end flat blade at one end and a larger round spoon at the other end 24.76cm Case of 100 Each for $595.41
14-245-98 Corning™
C3006
V-scoop at one end and a larger round spoon at the other end 24.9cm Case of 100 Each for $595.41
14-245-99 Corning™
C3007
Flat end blade at one end and a larger round spoon at the other end 24.76cm Case of 100 Each for $595.41
Specifications
Description
  • Single-use and disposable
  • RNase-/DNase-free
  • Antistatic, nonpyrogenic
  • Radation sterilized, individually packaged
  • Meets USP Class VI Requirements
Order Info
Individual packages have been radiation sterilized
Compliance
Nonpyrogenic certification
Meet USP Class VI Requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
USP Class VI, non-pyrogenic, RNase and DNase-free Certified
Yes
Polystyrene
Spatula
Molecular biology applications, lyophilized samples and contamination-free sampling
Yes