Please login to your online account to display your discounted pricing

Corning™ Square and Rectangular Cover Glasses


Provides quality, reliability and performance for your laboratory microscopy needs

Manufacturer: corning™  C2980244

 View more versions of this product

Catalog No. 12-553-470

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Material Borosilicate Glass
Shape Rectangular
Length (Metric) 40mm
Width (Metric) 24mm
Thickness (Metric) 0.16 to 0.19mm
Sterile No
For Use With Microscopes
Product Type Coverglass