Please login to your online account to display your discounted pricing
Corning™ Single-Use Bags for HYPERStack™ Vessels
Used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution
Manufacturer: corning™  9120077
 View more versions of this product
Catalog No. MT9120077
  • / Each
Description & Specifications
Specifications
Capacity (Metric) 20L
For Use With Hyperstack™ vessels to add media substrates to complete cell culture in an entirely closed solution
Quantity Each
Film Description ULDPE
Port Detail Port 1-12 L x 0.118 I.D. x 0.164 in. O.D. PVC tubing, 0.37 in. x 0.12 in. reducer, Port 2-male MPC connector with 0.37 in. barb female MPC cap, male luer connector with female luer cap, swabbable valve, bag port with 0.5 in. barb
Tubing Style Heat sealable flexible tubing