Please login to your online account to display your discounted pricing

Corning™ SFCA Syringe Filters


High burst strength for assured performance

Manufacturer: corning™  C431218

 View more versions of this product

Catalog No. 09-754-12

  • / Case of 50 Each
 

Description & Specifications

Specifications

Hold up Volume (After Air Purge) 100μL
Volume (Metric) 0.5 to 100mL
Pore Size 0.2μm
Tip Material Acrylic
Type Syringe Filter
Diameter (Metric) 26mm
Material Surfactant-free Cellulose Acetate (SFCA) w/prefilter membrane
Pressure Rating 87psi
Applications For tissue culture and HPLC