Please login to your online account to display your discounted pricing

Corning™ SF Medium


Based on a proprietary combination of classical media formulations

Manufacturer: corning™  40101CV

 View more versions of this product

Catalog No. MT40101CV

  • / Pack of 6 Each
 

Description & Specifications

Specifications

pH 7.2 to 7.6
Quantity 6 x 500mL
Storage Requirements 2° to 8°C
Product Type SF Medium, With L-Glutamine
Sterile Yes