Please login to your online account to display your discounted pricing

Corning™ Screw Caps for Disposable Culture Tubes


With white rubber liner

Manufacturer: corning™  C9999913

 View more versions of this product

Catalog No. C99999-13

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Color Black
Autoclavable Yes
Thread Size 13-415
Height (Metric) 13.5mm
Material Phenolic
For Use With (Equipment) Disposable culture tubes
Liner Material Rubber
Product Type Screw Cap

  • Made of a black phenolic compound
  • Disposable
  • Autoclavable