Corning™ rLaminin-521 (Human)
Recombinant heterotrimer protein composed of α5, β2, and γ1 chains
Manufacturer:  Corning™ C354223
 View more versions of this product
Catalog No. CB40223
Specifications
Description
Protein, Recombinant; Corning; rLaminin-521, Human; Heterotrimer composed of alpha5, beta2, and gamma1 chains expressed in mammalian cell culture system; Supports undifferentiated expansion of hPSC; Qty: 5mg
  • Expressed in mammalian cell culture system
  • Supports long-term self-renewal of human pluripotent stem cells (hPSC)
  • Provides ROCK inhibitor independent single cell expansion of PSCs, which improves hPSC culture ease and efficiency
  • Easy to self-coat
  • Available in 0.1mg aliquots for easy handling
  • Compatible with multiple hPSC culture media and dissociation reagents
  • Supports undifferentiated expansion of hPSC
  • Suitable for xeno-free, defined culture environment
Description & Specifications
Specifications
rLaminin-521, Human
5mg (50 x 100μg Vials)
Human
Yes