Please login to your online account to display your discounted pricing

Corning™ Reusable Polypropylene Low-Form Beakers

Translucent polypropylene beakers printed with blue graduations.

$44.20 - $49.95

Specifications

Autoclavable Yes
Certifications/Compliance DIN, ASTM Class A and B
Color Translucent
Material PP (Polypropylene)
Product Type Beaker
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    

S02291B

 
corning™
1000P-100
100mL Case of 12 Each for $44.20

S02291D

 
corning™
1000P-250
250mL Case of 6 Each for $49.95
Description & Specifications

Specifications

Autoclavable Yes
Certifications/Compliance DIN, ASTM Class A and B
Color Translucent
Material PP (Polypropylene)
Product Type Beaker
Disposable No
For Use With (Application) For high temperature and chemical resistance
Graduated Yes
Scale Blue graduated scale
Style Griffin
Temperature (English) Max. Operating 68°F
Temperature (Metric) Max. Operating 20°C