missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap
Translucent polypropylene; with polypropylene screw cap.
Manufacturer: Corning™ C1500P500
Description
These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.
Specifications
8.7 cm | |
Cylindrical | |
Translucent | |
10/Cs. | |
Non-sterile |
17.2 cm | |
500 mL | |
Polypropylene | |
1/Pk. |