missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap

Translucent polypropylene; with polypropylene screw cap.

Manufacturer:  Corning™ C1500P250

 View more versions of this product

Catalog No. 07-202-106



Description

Description

These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.

Specifications

Specifications

7.3 cm
Cylindrical
Translucent
20/Cs.
Non-sterile
13.2 cm
250 mL
Polypropylene
1/Pk.
SDS
Documents
Product Certifications