Please login to your online account to display your discounted pricing
PYREX™ Watch Glasses
Heavy walls and uniform fire-polished edges for strength.
$65.15 - $65.15
Specifications
Disposable No
Finish Plain
For Use With (Application) Used as beaker covers, permits escape of excess vapor and eliminates risk of contamination from dropped hooks or props
Material Pyrex™
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) For Use With (Equipment) Price Quantity    
S34821
fisher science education
9985100EMD
3.93 in. 10cm For 600mL beaker Pack of 12 Each for $65.15
Case of 12 Pack for $781.65
Description & Specifications
Specifications
Disposable No
Finish Plain
For Use With (Application) Used as beaker covers, permits escape of excess vapor and eliminates risk of contamination from dropped hooks or props
Material Pyrex™
Feature a high chemical durability which prevents contamination.