PYREX™ VISTA™ Round-Bottom Boiling Flasks
Designed for great shock resistance.
Manufacturer:  Fisher Science Education 70320A50E
 View more versions of this product
Catalog No. S76105F
Description
Description
  • Full length standard taper outer joints
  • Short necks
  • Manufactured from No. 7740 borosilicate glass
  • Wall thicknesses provide balance between thermal expansion and mechanical strength
Compliance
Designed to ASTM™ E-1403.
Specifications
Specifications
Education
50 mL
No
19/38
Round
Borosilicate Glass
Short
Documents