Please login to your online account to display your discounted pricing

PYREX™ VISTA™ Rimless Culture Tubes


Offer convenient plugging and rack storage.

Manufacturer: fisher science education  70820-25/EM

 View more versions of this product

Catalog No. S76111K

  • / Pack of 50 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Capacity (Metric) 55mL
Length (Metric) 150mm
Surface Untreated
Design Type Fire Polished
Disposable No
Rimmed No
Graduated No
For Use With (Application) For convenient plugging and rack storage
Closure Type Without
Material Borosilicate Glass
Shape Round
Label Type None
Diameter (Metric) Outer 25mm
Sterile No