PYREX™ VISTA™ Micro Filter Flask
Small flasks recommended for microchemical use.
Manufacturer:  Fisher Science Education 7036025EM
 View more versions of this product
Catalog No. S76107A
Description
Description
  • Permanent blue enamel marking spots
  • With tubulation
Compliance
Designed to ASTM™ E-1406, Type 2.
Specifications
Specifications
Education
1 cm
Borosilicate Glass
Narrow
Style: Filtering; Top style: Beaded rim
25 mL
No
Non-sterile
Documents