Please login to your online account to display your discounted pricing
PYREX™ VISTA™ Griffin Beakers
Provide balance between thermal shock resistance and mechanical strength.
$39.50 - $73.60
Specifications
Certifications/Compliance ASTM E960
Material Borosilicate Glass
Product Type Beaker
For Use With (Application) For optimum balance between thermal shock resistance and mechanical strength
Label Type Extra large marking
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    
S76100A
fisher science education
70000-10/EM
10mL Pack of 12 Each for $60.45
Case of 2 Pack for $120.85
S76100D
fisher science education
70000-50/EM
50mL Pack of 12 Each for $54.70
Case of 2 Pack for $109.35
S76100B
fisher science education
70000-20/EM
20mL Pack of 12 Each for $52.00
Case of 2 Pack for $103.95
S76100E
fisher science education
70000-100/E
100mL Pack of 12 Each for $57.75
Case of 2 Pack for $115.45
S76100F
fisher science education
70000-150/E
150mL Pack of 12 Each for $54.70
Case of 2 Pack for $109.35
S76100H
fisher science education
70000-400/E
400mL Pack of 12 Each for $62.80
Case of 2 Pack for $125.55
S76100K
fisher science education
70000-1L/EM
1000mL Pack of 6 Each for $73.60
Case of 2 Pack for $147.15
S76100G
fisher science education
70000-250/E
250mL Pack of 12 Each for $52.65
Case of 2 Pack for $105.30
S76100J
fisher science education
70000-600/E
600mL Pack of 6 Each for $39.50
Case of 2 Pack for $79.00
Description & Specifications
Specifications
Certifications/Compliance ASTM E960
Material Borosilicate Glass
Product Type Beaker
For Use With (Application) For optimum balance between thermal shock resistance and mechanical strength
Label Type Extra large marking
Style Griffin
  • Extra-large marking spot
  • 250 through 1000mL beakers have a double-graduated metric scale to indicate approximate content (10mL size is not graduated)
Designed to ASTM™ E-960.