PYREX™ VISTA™ Flat-Bottom Boiling Flasks
Designed for great shock resistance.
Manufacturer:  Fisher Scientific 701001LEM
 View more versions of this product
Catalog No. S76104D
Description
Description
  • Full length standard taper 24/40 outer joints
  • Short necks
  • Wall thicknesses provide balance between thermal expansion and mechanical strength
Compliance
Designed to ASTM™ E-1403.
Specifications
Specifications
Flat
Borosilicate Glass
Short
1000 mL
No
24/40
Documents