Please login to your online account to display your discounted pricing
PYREX™ VISTA™ Flat-Bottom Boiling Flasks
Designed for great shock resistance.
Manufacturer: fisher science education  70100-125/E
 View more versions of this product
Catalog No. S76104A
  • / Case of 12 Each
Description & Specifications
Specifications
Material Borosilicate Glass
Bottom Shape Flat
Capacity (Metric) 125mL
Disposable No
Neck Style Short
Standard Taper Joint 24/40