PYREX™ VISTA™ Class B Volumetric Flasks with Snap Caps
Capacity tolerances are twice those specified for Class A volumetric ware.
Manufacturer:  Fisher Scientific 7058110EMD
 View more versions of this product
Catalog No. S95185
Description
Description
  • Large, blue, block letters make inscriptions easy to read
  • Sharp, permanent graduation lines
  • Snap caps are supplied with all sizes
Specifications
Specifications
Flat
Blue
10 mL
Long
Calibrated To Contain
LDPE
Glass, Pyrex Glass
Documents