PYREX™ VISTA™ Class B Volumetric Flasks with Snap Caps
Capacity tolerances are twice those specified for Class A volumetric ware.
Manufacturer: Fisher Science Education 70581100EMD
Description
- Large, blue, block letters make inscriptions easy to read
- Sharp, permanent graduation lines
- Snap caps are supplied with all sizes
Specifications
Calibrated To Contain | |
LDPE | |
Glass,Pyrex Glass | |
Flat |
Blue | |
100 mL | |
Long |