Corning™ PYREX™ VISTA™ Class A Volumetric Flask
Necks are tooled for standard taper glass stoppers.
Supplier: Corning™ 70640500E
Description
- Provide precise volume measurement
- Graduation line is sharp and permanent
- Large blue block letters make the inscription easy to read
Compliance
Manufactured to Class A tolerances as established by ASTM™ E-694 for volumetric ware, ASTM E-542 for calibration of volumetric ware, and ASTM E-288 for volumetric flasks.
Specifications
Glass | |
Glass, Borosilicate Glass | |
Yes |
500 mL | |
Long | |
Flat |