missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PYREX™ VISTA™ Class A Cylinder
Calibrated “To Contain.”
Manufacturer: Fisher Science Education 7002210EM
Description
- Blue enamel graduations
- Capacity is in accordance with ASTM™ E-1272
- 10mL size has a funnel top for easier filling
Specifications
Graduated Cylinder | |
Blue | |
ASTM E1272 | |
Borosilicate Glass | |
Funnel Top, Pourout |
To Contain | |
Non-Sterile | |
A | |
10 mL | |
Single |