Please login to your online account to display your discounted pricing

PYREX™ Reagent Bottles with PYREX Hollow standard taper Stoppers


Plain, narrow mouth bottles with ground necks and hollow glass stoppers.

Manufacturer: fisher science education  15001LEMD

 View more versions of this product

Catalog No. S31224

  • / Pack
  • / Case of 24 Pack
 

Description & Specifications

Specifications

Height (Metric) 235mm
Capacity (English) 33.81 oz.
Diameter (English) 4.21 in.
Mouth Narrow
Sterile No
Capacity (Metric) 1000mL
Closure Hollow Glass Stopper
Diameter (English) Outer Neck 1.77 in.
Diameter (Metric) 107 mm
Shape Cylindrical
Capacity 33.81 oz., 1000mL
Height (English) 9.25 in.
Material Glass
Stopper Number 29
Diameter (Metric) Outer Neck 45mm