PYREX™ Griffin Beakers
With spouts and strengthened rim with flare trimmed back that offer optimum balance between thermal shock resistance and mechanical strength.
$77.00 - $121.33
Specifications
Material Borosilicate Glass
Disposable No
Graduated Yes
For Use With (Application) For Optimum Balance Between Thermal Shock Resistance and Mechanical Strength
Includes Spout
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S14269 View Documents Fisher Scientific
100030EMD
50 mL Pack of 12 Each for $101.85
Case of 4 Pack for $407.40
S14270 View Documents Fisher Scientific
100050EMD
100 mL Pack of 12 Each for $107.75
Case of 4 Pack for $431.00
S14271 View Documents Fisher Scientific
1000150EMD*
150 mL Pack of 12 Each for $97.78
Case of 4 Pack for $391.12
S14273 View Documents Fisher Scientific
1000400EMD*
400 mL Pack of 12 Each for $121.33
Case of 4 Pack for $485.32
S14272 View Documents Fisher Scientific
1000250EMD*
250 mL Pack of 12 Each for $103.23
Case of 4 Pack for $412.92
S14274 View Documents Fisher Scientific
1000600EMD*
600 mL Pack of 6 Each for $77.00
Case of 6 Pack for $462.00
Description
Description
  • With pouring spout
  • Extra-large marking spot
  • Dual white scale 5
  • 50mL sizes suitable for microanalysis
  • 250mL through 4000mL capacity beakers have double graduated metric scale
Compliance
Designed to meet ASTM™ E-960 requirements
Specifications
Specifications
Borosilicate Glass
Yes
Spout
ASTM E960
No
For Optimum Balance Between Thermal Shock Resistance and Mechanical Strength
Beaker
White Marking