missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Berzelius Tall-Form Beakers

A popular style for mixing in the lab.

Manufacturer:  Fisher Science Education 1060300EMD

 View more versions of this product

Catalog No. S63337




Description

Description

  • Taller than traditional Griffin style
  • Made with uniform wall thickness with extra-large enamel marking spot

Specifications

Specifications

ASTM E960
White Enamel Scale
10.14 oz.
6.4 cm
Borosilicate Glass
25 to 250 mL
11.8 cm
25 mL
12 Pk.
Yes
Berzelius
2.51 in.
300 mL
Convenient for Easy Pouring
4.64 in.
Spout
Extra Large Enamel Marking
Beaker
SDS
Documents
Product Certifications