Please login to your online account to display your discounted pricing
Corning™ PureCoat™ Carboxyl Surface Dishes
Negatively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
$163.23 - $406.05
Specifications
Surface Coating Carboxyl
View More Specs
 Alert
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Products
Catalog Number Mfr. No. Diameter (Metric) Price Quantity    
08-772-168
corning™
C354784
100mm; 5 packs; Case of 5 Case of 10 Each for $163.23
08-772-178
corning™
C356784
100mm; 5 packs; case of 50 Case of 40 Each for $406.05
Description & Specifications
Specifications
Surface Coating Carboxyl
  • Chemically defined, animal-free
  • Enhances recovery from freeze-thaw
  • Suitable for primary, transfected and fastidious cell lines
  • Maintains cell monolayers during vigorous liquid handling manipulation