Please login to your online account to display your discounted pricing

Corning™ PureCoat™ Carboxyl Rectangular Canted Neck Cell Culture Flasks with Vented Cap


Negatively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays

Manufacturer: corning™  C354780

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-026

  • / Case of 5 Each
 

Description & Specifications

Specifications

Neck Style Canted
Capacity (English) 25.3 oz.
Capacity (Metric) 750mL
Culture Area 175cm2
Closure Type Vented
Surface Coating Carboxyl
Material Polystyrene
Capacity 25.3 oz., 750mL
For Use With (Application) For preparative cell culture, cell-based and HTS screening assays
Shape Rectangular